Learn More
Bio Rad Laboratories PREPARATIVE COMB FOR SUB-CELL
Supplier:  Bio Rad Laboratories 1704328
Encompass
Comb, Preparative; Bio-Rad; For Sub-Cell, Wide Mini-Sub Cell Systems, 1 preparative, 2 reference wells, 3mm thick, adjustable height
Catalog No. 50-901-1139