Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Prepared Bottled Media: Rinsing Fluid D

Catalog No. 22349720 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Rinsing Fluid D; 300mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-349-720 Rinsing Fluid D; 300mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-349-720 Supplier BD Supplier No. 257536

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For detection of microorganisms from pharmaceutical, food, environmental, or water testing samples

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Rinsing Fluid D
For Use With (Application) Detection of microorganisms from pharmaceutical, food, environmental or Water testing samples
Packaging Type Bottle
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.