Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Prepared Bottled Media: Tryptic Soy Broth, Wide Mouth Bottle

Catalog No. 22349999 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
90 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-349-999 90 mL
22-349-998 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-349-999 Supplier BD Supplier No. 257488
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples

  • Ready for immediate use
  • Sterility Assurance Level of 10-6 reduces risk of false positives
  • Terminally autoclaved
  • Conforms to Harmonized USP, EP, and JP requirements for sterility tests and to CLSI requirements for recovery

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Tryptic Soy Broth
Cap Type Lug cap
Certifications/Compliance USP
Color Amber
Country of Origin United States
Detects Microorganisms
Form Liquid
Format Bottle
Manufacturing Origin United States
Media Type Tryptic Soy Broth
Product Type Broth
Sterility Sterile
Test Type Microbial Detection
Fill Volume 90 mL
Final Product Type Microbial Growth
Incubation Conditions 30°C to 35°C
Incubation Time 24 to 48 hr.
Manual/Automated Manual
Packaging Type Bottle
Product Line BD Difco
Quantity 90 mL
Specimen Type Pharmaceutical Samples, Food Samples, Environmental Samples, Water Samples
Width (English) 1.89 in.
Width (Metric) 48 mm
Height (Metric) 130 mm
Diameter (English) Outside 1.89 in.
Diameter (Metric) 48 mm
Protein Source Soybean
Manufacturing Quality EP/USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.