Learn More
Description
- Each bottle is labeled with product name, expiration date and lot number for easy handling
- Bottles are narrow-mouth with black phenolic screw caps
- Capacity: 125mL each
Order Info
Sold in packs of 24 bottles.
Specifications
Specifications
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Nutrient Broth |
| Product Type | Prepared Microbiology Media |
| Sterility | Sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbial Identification |
| Fill Volume | 125 mL |
| Packaging Type | Bottle |
| Quantity | 125 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
