Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Prepared Culture Media in Tubes
SCI_ED

Catalog No. S716992A Shop All Fischer Technical Company Products
Change view
Click to view available options
Fill Volume:
5 mL
10 mL
Media Type:
BHI Broth
Nutrient Broth
SIM Medium
Tryptic Soy Broth
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume Media Type
S716992A 10 mL Tryptic Soy Broth
S716942A 10 mL Nutrient Broth
S012394 5 mL SIM Medium
S012397 10 mL BHI Broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Prop 65: Cancer and Reproductive Harm.
Catalog No. S716992A Supplier Fischer Technical Company™ Supplier No. CM164T1020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Prop 65: Cancer and Reproductive Harm.

Ready-to-use media prepared in disposable borosilicate glass tubes.

  • Tubes measure 16 mm O.D. x 125 mm long
  • Include black plastic screw caps
  • Each tube is labeled with product name, expiration date and lot number
  • Refrigerate until ready to use

Order Info

Sold in pack of 20 tubes.

Specifications

Manufacturing Origin US with Globally Sourced Materials
Media Type Tryptic Soy Broth
Product Type Prepared Microbiology Tubes
Sterility Sterile
Target Organism Class Bacteria
Test Type Microbial Identification
Fill Volume 10 mL
Quantity 20 Tubes/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.