Learn More
Braintree Scientific Prepared MicroTube w/Screw Cap, 1.3ml, Serum/Clotting Activa
SDP
Catalog No. 501955254
Encompass
Prepared Micro Tubes with Screw Cap Closure. Closure contains integral O-ring. Sample volume 1.3ml. Skirted, conical base and Label **Sold for research purposes only and not for human use**
Catalog No. 50-195-5254 Supplier Braintree Scientific Supplier No. MT1392
Prepared Micro Tubes with Screw Cap Closure. Closure contains integral O-ring. Sample volume 1.3ml. Skirted, conical base and Label **Sold for research purposes only and not for human use**