Learn More
Braintree Scientific Prepared Microtubes with Push Cap,1.3ml,Lithium Heparin
SDP
Catalog No. 501955528
Encompass
Prepared Micro Tubes with Attached Push Cap Closure. Skirted, conical base and label. Preparations: Serum clotting activator, plasma lithium heparin, hematology potassium EDTA and coagulation citrate **Sold for research purposes only and not for human use**
Catalog No. 50-195-5528 Supplier Braintree Scientific Supplier No. PMT1503CS
Prepared Micro Tubes with Attached Push Cap Closure. Skirted, conical base and label. Preparations: Serum clotting activator, plasma lithium heparin, hematology potassium EDTA and coagulation citrate **Sold for research purposes only and not for human use**