Learn More
Braintree Scientific Prepared Microtubes with Push Cap,1.3ml, EDTA, 1,000/cs
SDP
Catalog No. 501955530
Encompass
Prepared Micro Tubes with Attached Push Cap Closure. Skirted, conical base and label. Preparations: Serum clotting activator, plasma lithium heparin, hematology potassium EDTA and coagulation citrate **Sold for research purposes only and not for human use**
Catalog No. 50-195-5530 Supplier Braintree Scientific Supplier No. PMT1504CS
Prepared Micro Tubes with Attached Push Cap Closure. Skirted, conical base and label. Preparations: Serum clotting activator, plasma lithium heparin, hematology potassium EDTA and coagulation citrate **Sold for research purposes only and not for human use**