Learn More
Wards Natural Science Est Stentor Slide
Catalog No. NC1621634
Encompass_Preferred
Ciliate protist species, identifying features clearly distinguishable rigorous quality control standards. Trumpet-shaped spirotrich. Large macro-nucleus appears in the form of a long chain of beads.
Catalog No. NC1621634 Supplier Wards Natural Science Est Supplier No. 470177504
Ciliate protist species, identifying features clearly distinguishable rigorous quality control standards. Trumpet-shaped spirotrich. Large macro-nucleus appears in the form of a long chain of beads.