Learn More
Waters Corp Waters Corporation PrepLC Module, 25 mm
Supplier:  Waters Corp WAT015814
Encompass_Preferred
The PrepLC, 25 mm Module is a versatile and durable preparative liquid chromatography module designed for the efficient separation and purification of large volumes of samples in various applications. It features a 25 mm inner diameter column for high flow rates and excellent separation efficiency, ensuring reliable and precise results. It is compatible with a wide range of mobile phases and can be seamlessly integrated into existing laboratory workflows, being compatible with various lab equipment and software packages.
  • High throughput for purifying large sample volumes.
  • Excellent separation efficiency for reliable results.
  • User-friendly design and easy maintenance.
  • Versatile for diverse research needs.
  • Features a 25 mm inner diameter column.
  • Compatible with a wide range of mobile phases and lab equipment.
  • Crafted from high-quality materials for durability.
Catalog No. 50-785-080