Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ PrepMan™ Ultra Sample Preparation Reagent
GREENER_CHOICE

Catalog No. 4318930
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-189-30 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-189-30 Supplier Applied Biosystems™ Supplier No. 4318930
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The PrepMan™ Ultra Sample Preparation Reagent was developed for the rapid preparation of DNA template from Gram-negative food-borne pathogens for use in PCR amplification reactions.

The PrepMan™ Ultra Sample Preparation Reagent was developed for the rapid preparation of DNA template from Gram-negative food-borne pathogens for use in PCR amplification reactions. These samples often have high lipid content that can inhibit PCR amplification of the template. Using a simple boil and spin protocol, PrepMan™Ultra Reagent efficiently inactivates PCR inhibitors, significantly reducing the need to repeat the template preparation step.

PrepMan™ Ultra reagent has been used to prepare PCR-ready template from a wide variety of different sources, including bacteria, yeast, filamentous fungi, both from a plate or from liquid culture, mammalian tissue smears, human cells (buccal swab), and mammalian whole blood. The resulting template has been used amplified in allelic discrimination assays, genetically modified organism (GMO) detection and quantification, food pathogen detection, microbial identification, and bacterial strain typing.

  • Simple, rapid protocol reduces time to results.
  • Unique formulation inactivates PCR inhibitors, yielding consistent results.
  • PCR-ready template can be prepared from a wide range of sample types.
  • Affordable method with a low price per preparation.
  • Homogeneous solution with minimal sample-handling requirements.
  • Microcentrifuge tube-based format means no additional equipment is needed for template preparation.

PrepMan™ Ultra reagent is a novel formulation and is a homogeneous solution that does not contain Chelex™ or any other type of resin or matrix.

This product is a replacement⁄improvement of PrepMan™ Sample Preparation Reagent, and should be recommended to current and potential PrepMan™ Reagent users.

Used With
3500/3500xL Genetic Analyzer Capillary Array (50 cm), 3130xL/3100 Genetic Analyzer Capillary Array (36 cm), 96-Well Plate Septa, AmpliTaq Gold™ PCR Master Mix, BigDye™ Terminator v1.1 Cycle Sequencing Kit, POP-6™ Polymer for 3100/3100-Avant™ Genetic Analyzers, PrepMan™ Ultra Sample Preparation Reagent, TaqMan™ GMO Maize 35S Detection Kit, TaqMan™ GMO Soy 35S Detection Kit.

Specifications

Content And Storage Contents: One bottle of reagent.

Store the PrepMan Ultra Sample Preparation Reagent, at room temperature. Do not freeze or autoclave the PrepMan Ultra Sample Preparation Reagent.
Format Bottle
Sample Type Bacterial Culture, Food, Fungi, Whole Blood
Final Product Type Genomic DNA, Bacterial DNA
For Use With (Application) Real-Time Quantitative PCR (qPCR), Sequencing, PCR
No. of Reactions 200 Preps
Product Line PrepMan
Quantity 1 kit
Shipping Condition Room Temperature
System Type MicroSEQ™ ID
Test Time 15 min
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.