Learn More
Waters Corp Porasil Silica Radial-Pak Column, 125Å, 40 µm, 47 mm X 50 mm, 10/pk
Catalog No. 50785248 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Porasil 15 - 20µm silica packing material provides long column life and low backpressure. The packing also has a high surface area making it ideal for preparative work.
Catalog No. 50-785-248 Supplier Waters Corp Supplier No. WAT025877
Add to Cart
Add to Cart
Request a Quote
Waters Porasil 15 - 20µm silica packing material provides long column life and low backpressure. The packing also has a high surface area making it ideal for preparative work.