Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI PrepPure prep HPLC Columns - ID 30
SDP

Catalog No. p-7213534 Shop All BUCHI Products
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
Size (Metric) Tube Rack:
10 μm
15 μm
5 μm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Size (Metric) Tube Rack
05-405-502 150 mm 5 μm
05-405-503 250 mm 5 μm
05-405-514 150 mm 10 μm
05-405-515 250 mm 10 μm
05-405-526 150 mm 15 μm
05-405-527 250 mm 15 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 05-405-502 Supplier BUCHI Supplier No. 11068667
Only null left
Add to Cart
Add to Cart
Request a Quote

PrepPure HPLC columns are filled with high quality silica and enable the performance of high resolution separations.

  • Wide range of silica phases (unbonded Silica, C18, C18 AQ, C4) and particle sizes (5-15μm)
  • Easy scalability thanks to multiple dimensions (4.6-70mm ID; 150/ 250mm lengths)
  • C18 is the most popular nonpolar phase for prep HPLC separations
  • C18 AQ is stabilized for polar to medium polar compounds in a high water environment
  • C4 is designed for high molecular weight compounds (>2000 Da) like peptides and proteins
  • Silica is used for high and medium polar compounds and organic solvents
  • 70mm ID columns for large scale purification (up to 85g for Silica and 8.5g for C18)

Specifications

Length (Metric) 150 mm
Type C18
Size (Metric) Tube Rack 5 μm
Column Type HPLC
Max. Pressure 210 bar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.