Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caresfield Preprinted Label
SDP

Catalog No. 22246085
Change view
Click to view available options
Display Text:
ABO and Rh Confirmed
Name Alert
Pre-Filtered
Received On
Serum
Sickle Screen Negative
Special Antigen Typing
Store at Room Temperature
This Lot is Ready for Use
UN3373 Biological Substance
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
22-246-085 Sickle Screen Negative
22-246-075 Serum
22-246-074 Name Alert
22-246-082 Pre-Filtered
22-246-083 Received On
22-246-076 This Lot is Ready for Use
22-246-077 Store at Room Temperature
22-246-080 ABO and Rh Confirmed
22-246-081 Special Antigen Typing
22-246-084 UN3373 Biological Substance
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 22-246-085 Supplier Caresfield Supplier No. 16967
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Preprinted alert labels for various applications.

  • Preprinted
  • Identification label
  • Bright colors for enhanced visibility

Specifications

Autoclavable Not Autoclavable
Color Capability Multicolor
Adhesive Type Acrylic
Coating None
Color Blue
Finish Matte
Format Roll
Print Technology None
Product Type Custom Labels
Sterility Non-sterile
Display Text Sickle Screen Negative
Languages English
Latex Free Yes
No. per Pack 500/Roll, 6 Roll/Cs.
Resolution 203 dpi
Shape Rectangle with Rounded Corners
Dimensions (L x W) 1.25 x 0.3125 in.
Length (English) 1.25 in.
Width (English) 0.3125 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.