Learn More
Research Products International Corp PrepSafe Floating Micro-Tube Rack
SDP
Catalog No. 501978549
Encompass
PrepSafe Micro-Tube Rack features a unique design that holds tubes secure in place and prevents caps from opening during heating or boiling applications. Accommodates up to twenty eight 1.5-2.0ml flip top micro-tubes. Each location is numbered to allow quick identification of samples. Ideal for use in water baths and ice buckets. Floating rack keeps tube lids above water and prevents sample contamination. Rack is autoclavable.
Catalog No. 50-197-8549 Supplier Research Products International Corp Supplier No. 246725
PrepSafe Micro-Tube Rack features a unique design that holds tubes secure in place and prevents caps from opening during heating or boiling applications. Accommodates up to twenty eight 1.5-2.0ml flip top micro-tubes. Each location is numbered to allow quick identification of samples. Ideal for use in water baths and ice buckets. Floating rack keeps tube lids above water and prevents sample contamination. Rack is autoclavable.