Learn More
Research Products International Corp PrepSafe Floating Micro-Tube Rack, Yellow
SDP
Catalog No. 501367893
Encompass
PrepSafe Racks protect samples from loss or contamination during incubation procedures or when chilled in an ice bucket. Unique locking tabs hold tubes securely in place and prevent lids from popping open during boiling applications. Racks are designed to float in a water bath or most 1000ml beakers. Manufactured from polypropylene and can be autoclaved. Each rack holds up to twelve 1.5 - 2.0 ml micro-tubes.
Catalog No. 50-136-7893 Supplier Research Products International Corp Supplier No. 246726Y
PrepSafe Racks protect samples from loss or contamination during incubation procedures or when chilled in an ice bucket. Unique locking tabs hold tubes securely in place and prevent lids from popping open during boiling applications. Racks are designed to float in a water bath or most 1000ml beakers. Manufactured from polypropylene and can be autoclaved. Each rack holds up to twelve 1.5 - 2.0 ml micro-tubes.