Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC preQ1 dihydrochlorid 100mg | 86694-45-3 | 252.10 g/mol | C7H11Cl2N5O | 100 MG
SDP

Catalog No. 5000229389
Encompass_Preferred

preQ1 dihydrochloride is the dihydrochloride salt of pre-queuosine 1, a guanine-derived nucleobase that serves as a direct precursor in queuine biosynthesis. It binds with high affinity to the PreQ1 riboswitch aptamer and is provided for biochemical and molecular biology research (research use only).

  • Precursor in queuine biosynthesis and riboswitch studies.
  • Molecular formula C7H11Cl2N5O; molecular weight 252.10 g/mol.
  • Purity 98.88% as supplied.
  • Soluble in DMSO (125 mg/mL) and water (14.29 mg/mL with warming).
  • Appearance solid, white to light brown.
  • Available in small research quantities (1 mg-500 mg).

Catalog No. 50-002-29389 Supplier Medchemexpress LLC Supplier No. HY115642100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

preQ1 dihydrochloride is the dihydrochloride salt of pre-queuosine 1, a guanine-derived nucleobase that serves as a direct precursor in queuine biosynthesis. It binds with high affinity to the PreQ1 riboswitch aptamer and is provided for biochemical and molecular biology research (research use only).

  • Precursor in queuine biosynthesis and riboswitch studies.
  • Molecular formula C7H11Cl2N5O; molecular weight 252.10 g/mol.
  • Purity 98.88% as supplied.
  • Soluble in DMSO (125 mg/mL) and water (14.29 mg/mL with warming).
  • Appearance solid, white to light brown.
  • Available in small research quantities (1 mg-500 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.