Learn More
Medchemexpress LLC preQ1 dihydrochlorid 10mg | 86694-45-3 | 252.10 | C7H11Cl2N5O | 10 MG
SDP
Catalog No. 5000228032
Encompass_Preferred
preQ1 dihydrochloride is a guanine-derived nucleobase used as a precursor in the biosynthesis of queuine and as a ligand for PreQ1 riboswitch studies. Supplied as the dihydrochloride salt for research use, the compound has defined solubility and storage recommendations.
  • Precursor in queuine biosynthesis and useful for tRNA modification studies.
  • Binds the PreQ1 riboswitch aptamer with high affinity.
  • Solubility: DMSO 125 mg/mL; water 15 mg/mL (may require ultrasonic agitation).
  • Solid storage recommended at 4°C in a sealed container, protected from moisture and light.
  • In-solution stability: store at -80°C for up to 6 months or -20°C for up to 1 month when sealed.
Catalog No. 50-002-28032 Supplier Medchemexpress LLC Supplier No. HY11564210MG
preQ1 dihydrochloride is a guanine-derived nucleobase used as a precursor in the biosynthesis of queuine and as a ligand for PreQ1 riboswitch studies. Supplied as the dihydrochloride salt for research use, the compound has defined solubility and storage recommendations.
  • Precursor in queuine biosynthesis and useful for tRNA modification studies.
  • Binds the PreQ1 riboswitch aptamer with high affinity.
  • Solubility: DMSO 125 mg/mL; water 15 mg/mL (may require ultrasonic agitation).
  • Solid storage recommended at 4°C in a sealed container, protected from moisture and light.
  • In-solution stability: store at -80°C for up to 6 months or -20°C for up to 1 month when sealed.