Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC preQ1 dihydrochlorid 1mg | 86694-45-3 | 252.10 g·mol⁻1 | C7H11Cl2N5O | 1 MG
SDP

Catalog No. 5000226269
Encompass_Preferred

PreQ1 dihydrochloride is a guanine-derived nucleobase precursor used in research on tRNA modification and queuine biosynthesis. It binds preQ1 riboswitch aptamers with high affinity and is supplied as the dihydrochloride salt for in vitro biochemical and molecular biology studies.

  • High purity, typically 98.88%.
  • Molecular weight 252.10 g·mol⁻1; formula C7H11Cl2N5O.
  • Soluble in DMSO (125 mg/mL) and water (≈14.29 mg/mL) with sonication and warming to 60°C.
  • White to light brown solid appearance.
  • Store sealed at 4°C, protect from moisture and light; in solution store at -80°C up to 6 months.

Catalog No. 50-002-26269 Supplier Medchemexpress LLC Supplier No. HY1156421MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PreQ1 dihydrochloride is a guanine-derived nucleobase precursor used in research on tRNA modification and queuine biosynthesis. It binds preQ1 riboswitch aptamers with high affinity and is supplied as the dihydrochloride salt for in vitro biochemical and molecular biology studies.

  • High purity, typically 98.88%.
  • Molecular weight 252.10 g·mol⁻1; formula C7H11Cl2N5O.
  • Soluble in DMSO (125 mg/mL) and water (≈14.29 mg/mL) with sonication and warming to 60°C.
  • White to light brown solid appearance.
  • Store sealed at 4°C, protect from moisture and light; in solution store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.