Learn More
Medchemexpress LLC preQ1 dihydrochlorid 25mg | 86694-45-3 | 252.10 g/mol | C7H11Cl2N5O | 25 MG
SDP
Catalog No. 5000226270
Encompass_Preferred
preQ1 dihydrochloride is the dihydrochloride salt of pre-queuosine1, a guanine-derived nucleobase and a direct precursor in queuine biosynthesis. It is supplied for research use as a biochemical probe that binds with high affinity to the PreQ1 riboswitch aptamer and is suitable for studies of tRNA modification and riboswitch function.
  • High purity suitable for research use (98.88%).
  • Solid, white to light brown appearance for easy handling.
  • Soluble in DMSO (125 mg/mL) and water (14.29 mg/mL) with sonication or warming.
  • Direct precursor in queuine biosynthesis and riboswitch-binding probe.
  • Stable when stored sealed away from moisture and light; in solvent -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-26270 Supplier Medchemexpress LLC Supplier No. HY11564225MG
preQ1 dihydrochloride is the dihydrochloride salt of pre-queuosine1, a guanine-derived nucleobase and a direct precursor in queuine biosynthesis. It is supplied for research use as a biochemical probe that binds with high affinity to the PreQ1 riboswitch aptamer and is suitable for studies of tRNA modification and riboswitch function.
  • High purity suitable for research use (98.88%).
  • Solid, white to light brown appearance for easy handling.
  • Soluble in DMSO (125 mg/mL) and water (14.29 mg/mL) with sonication or warming.
  • Direct precursor in queuine biosynthesis and riboswitch-binding probe.
  • Stable when stored sealed away from moisture and light; in solvent -80°C (6 months), -20°C (1 month).