Learn More
TARGETMOL CHEMICALS INC Preq1-Dihydrochloride 10MG
SDP
Catalog No. 502876026
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Preq1-Dihydrochloride (pre-queuosine1 Dihydrochloride) is an intermediate of the queuosine pathway, it binds with high affinity to the aptamer of PreQ1 riboswitch, which attenuates protein expression. Purity 97.14%
Catalog No. 50-287-6026 Supplier TARGETMOL CHEMICALS INC Supplier No. T901210MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Preq1-Dihydrochloride (pre-queuosine1 Dihydrochloride) is an intermediate of the queuosine pathway, it binds with high affinity to the aptamer of PreQ1 riboswitch, which attenuates protein expression. Purity 97.14%