Learn More
Cayman Chemical preQ1hydrochlorIde 10mg
SDP
Catalog No. 502964064
Encompass_Preferred
A precursor of queuosine; binds to an aptamer domain on tRNA that acts as a riboswitch to repress gene expression
Catalog No. 50-296-4064 Supplier Cayman Chemical Supplier No. 2186410 MG
A precursor of queuosine; binds to an aptamer domain on tRNA that acts as a riboswitch to repress gene expression