Learn More
Thomas Scientific Presat Wipers, Texwipe, TX1041 TechniSat 70% IPA Nonwoven Polycellulose Fabric, 70 per recloseable bag, 12 bags/case
SDP
Catalog No. NC1800950
Encompass
TechniSat incorporates a pre-wetted fabric blend of nonwoven hydroentangled 55% cellulose and 45% polyester. This fabric combines the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic fiber. It offers low particle counts, low extractable levels and solvent compatibility. TechniSat wipers are pre-wetted with isopropyl alcohol/deionized water solutions filtered to 0.2 µm. The wipers are packaged in resealable flexpacks, fliptop tubs and canisters. TechniSat containers are sheathed in a sealed outer bag for proper introduction into the cleanroom.
This item is not returnable. View return policy
Catalog No. NC1800950 Supplier Thomas Scientific Supplier No. TX1041
TechniSat incorporates a pre-wetted fabric blend of nonwoven hydroentangled 55% cellulose and 45% polyester. This fabric combines the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic fiber. It offers low particle counts, low extractable levels and solvent compatibility. TechniSat wipers are pre-wetted with isopropyl alcohol/deionized water solutions filtered to 0.2 µm. The wipers are packaged in resealable flexpacks, fliptop tubs and canisters. TechniSat containers are sheathed in a sealed outer bag for proper introduction into the cleanroom.