Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Preschool Paper Pack
SDPSCI_ED

Catalog No. S13202
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13202 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13202 Supplier Roylco™ Supplier No. R15325
Add to Cart
Add to Cart

Teach preschoolers how to cut, fold, collage and craft with these vibrant craft papers.

  • Beautiful mix of colors and patterns is a nice addition to preschool classroom
  • Introduce colors, patterns and animals with a wide range of delightful paper designs
  • Big selection of paper is packed with value
  • Sheets come in two sizes: 8.5 x 11 in. (22 x 28 cm) and 8 x 9 in. (20 x 23 cm)

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Format Kit
Includes Sheets, Guide
No. of Pages 176 Pages
Languages English, Spanish, French, German
Subject General Education
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.