Learn More
THORLABS INC Precision Cover Glasses, #1.5H Thickness, 22 x 22 mm, Pack of 200
Catalog No. NC1776158
Encompass
Precision Glass Cover Slips with #1.5H (170 µm) Thickness and ± 5 µm ToleranceRecommended for Use with High NA Objective LensesSquare (22 mm x 22 mm),Thorlabs' Precision Cover Glasses are ground to a uniform thickness of 170 ± 5 µm and are made from Schott D 263 M Glass, an optically clear and colorless glass that exhibits low auto-fluorescence. The uniform thickness and consistent optical properties help reduce image defects due to spherical aberrations (see the Specs tab for details). As such, these cover slips are ideal for use with high NA objective lenses.
Catalog No. NC1776158 Supplier THORLABS INC Supplier No. CG15CH2
Precision Glass Cover Slips with #1.5H (170 µm) Thickness and ± 5 µm ToleranceRecommended for Use with High NA Objective LensesSquare (22 mm x 22 mm),Thorlabs' Precision Cover Glasses are ground to a uniform thickness of 170 ± 5 µm and are made from Schott D 263 M Glass, an optically clear and colorless glass that exhibits low auto-fluorescence. The uniform thickness and consistent optical properties help reduce image defects due to spherical aberrations (see the Specs tab for details). As such, these cover slips are ideal for use with high NA objective lenses.