Learn More
Sigma Aldrich Fine Chemicals Biosciences PreScission Protease Cytiv
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE27084301
Encompass
PreScission Protease is a genetically engineered fusion protein of human rhinovirus 3C protease and glutothione S trasferase (GST). PreScisson protease specifically cleaves between the Gln and Gly residues of the recognition sequence LeuGluValLeuPheGln/GlyPro.
Catalog No. 50-290-4487