Learn More
Genscript Corporation PreScission Protease
Supplier:  Genscript Corporation Z02799100
Encompass
PreScission Protease is a fusion protein of glutathione S-transferase (GST) and human rhinovirus (HRV) type 14 3C protease. The optimum recognition site for this enzyme is the sequence Leu-Glu-Val-Leu-Phe-Gln/Gly-Pro (LEVLFQ/GP) and cleavage occurs between the Gln and Gly-Pro residues. Substrate recognition and cleavage are likely to be dependent not only upon primary structural signals, but also upon the secondary and tertiary structures of the fusion protein as well.
Catalog No. NC2124297