Learn More
Apexbio Technology LLC PreScission Protease (PSP), 500U (1U/uL). Cas: MFCD: NA. Specifically recognize and cleave the Leu-Glu-Val-Leu-Phe-Gln-Gly-Pro peptide in fusion proteins.
SDP
Catalog No. 501998219
Encompass_Preferred
PreScission Protease is a fusion protein of human rhinovirus 3C protease and glutothione S trasferase (GST). PreScisson protease can specifically recognize the Leu-Glu-Val-Leu-Phe-Gln/Gly-Pro sequence and cleaves between the Gln and Gly residues. Concentration: 2U/L Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8219 Supplier Apexbio Technology LLC Supplier No. K1101500
PreScission Protease is a fusion protein of human rhinovirus 3C protease and glutothione S trasferase (GST). PreScisson protease can specifically recognize the Leu-Glu-Val-Leu-Phe-Gln/Gly-Pro sequence and cleaves between the Gln and Gly residues. Concentration: 2U/L Other sizes are also available. Please inqury us for quote.