Learn More
Sigma Aldrich Fine Chemicals Biosciences Prescored ampules clear gl
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1849851PAK
Encompass
Stems can be pull or tip sealed to effect a hermetically sealed container. Made from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I Class A requirements as defined by the Pharmacopeia of the United States. Pre-scored to eliminate filing.
Catalog No. 50-289-9998