Learn More
NEOGEN Presence Absence Broth, 5 kg
SDP
Catalog No. 502015505
Encompass_Preferred
Presence Absence Broth is used for the detection of coliform bacteria in water treatment plants or distribution systems using the presence-absence coliform test in a laboratory setting. Presence Absence Broth is not intended for use in the diagnosis of disease or other conditions in humans. The Presence Absence (PA) test is a presumptive detection for coliforms in water. The test is a simple modification of the multiple-tube procedure. One 100 mL test sample is inoculated into a single culture bottle to obtain qualitative information on the presence or absence of coliforms, through the presence or absence of lactose fermentation. This test is based on the principle that coliforms and other pollution indicator organisms should not be present in a 100 mL water sample.For more information visit https://www.neogen.com/solutions/Microbiology/presence-absense-broth
Catalog No. 50-201-5505 Supplier NEOGEN Supplier No. 700003321
Presence Absence Broth is used for the detection of coliform bacteria in water treatment plants or distribution systems using the presence-absence coliform test in a laboratory setting. Presence Absence Broth is not intended for use in the diagnosis of disease or other conditions in humans. The Presence Absence (PA) test is a presumptive detection for coliforms in water. The test is a simple modification of the multiple-tube procedure. One 100 mL test sample is inoculated into a single culture bottle to obtain qualitative information on the presence or absence of coliforms, through the presence or absence of lactose fermentation. This test is based on the principle that coliforms and other pollution indicator organisms should not be present in a 100 mL water sample.For more information visit https://www.neogen.com/solutions/Microbiology/presence-absense-broth