Learn More
Hach Company Presence-Absence Broth Ampules, pk/25
Supplier:  Hach Company 2494925
Encompass
Presence/Absence Broth ampules provide an easy-to-use test, ideal for screening drinking water samples for the presence of total coliform bacteria. Simply combine the P/A broth with your 100mL sample and incubate for 24 to 48 hours at 35 degrees Celcius. A color change to yellow or yellow brown indicates presence of coliform bacteria.
Catalog No. 50-199-6074