Learn More
Enzo Life Sciences Presenilin 1 monoclonal antibody (APS 11) (200µg)
SDP
Supplier:  Enzo Life Sciences ALX804554C200
Encompass_Preferred
Clone: APS 11. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 21-34 (CH21LSNTVRSQND NRE34) of N-terminal human presenilin 1. Species reactivity: Human, Mouse, Rat Primate. Applications: ELISA, ICC, IHC (PS), WB. Purity: Protein G-affinity  purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-9944