Learn More
Enzo Life Sciences Presenilin 1 monoclonal antibody (APS 18) (200µg)
SDP
Supplier:  Enzo Life Sciences ALX804555C200
Encompass_Preferred
Clone: APS 18. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 313-334 (S313KYNAESTER ESQDTVAENDDG334) of C-terminal human presenilin 1. Species reactivity: Human, Mouse, Rat, Monkey. Applications: ELISA, ICC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-9945