Learn More
MSC Karcher Pre-separator Lid, For Industrial Vacuum, 1 ea
Catalog No. 502816433
Encompass
The Karcher pre-separator lid is a repair part designed for use with industrial vacuum systems. It functions as a pre-separator, collecting bulk dry materials before they reach the main vacuum filter, thereby protecting the vacuum and extending filter life.
  • Part type: Preseparator lid
  • For use with industrial vacuum
Catalog No. 50-281-6433 Supplier MSC Supplier No. 16617110
The Karcher pre-separator lid is a repair part designed for use with industrial vacuum systems. It functions as a pre-separator, collecting bulk dry materials before they reach the main vacuum filter, thereby protecting the vacuum and extending filter life.
  • Part type: Preseparator lid
  • For use with industrial vacuum