Learn More
Biosure Inc PRESERVATIVE FREE SHEATH
Catalog No. NC9867260
Encompass
BioSure Preservative-Free Sheath Solution is a buffered, support/carrier reagent for transporting particles through a flow cytometer. The product contains no antibacterial/antifungal preservative.
Catalog No. NC9867260 Supplier Biosure Inc Supplier No. 1020
BioSure Preservative-Free Sheath Solution is a buffered, support/carrier reagent for transporting particles through a flow cytometer. The product contains no antibacterial/antifungal preservative.