Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Pre-sharpened Double-Pointed Carbon Rod, 2" OAL, ¼" diameter
SDP

Catalog No. 501901838
Encompass_Preferred

All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite. Neck type tips.
OAL: 2"
Diameter: ¼"
Tip diameter: 0.039"
Tip: neck

Catalog No. 50-190-1838 Supplier Electron Microscopy Sciences Supplier No. 7022140
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite. Neck type tips.
OAL: 2"
Diameter: ¼"
Tip diameter: 0.039"
Tip: neck

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.