Learn More
Electron Microscopy Sciences Pre-sharpened Double Tip Carbon Rod, 2 3/8" OAL, ⅛" diameter
SDP
Catalog No. 5024908
Encompass_Preferred
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite.
OAL: 2 3/8"
Diameter: ⅛"
Tip diameter: 0.354"
Tip: 3/16"
Catalog No. 50-249-08 Supplier Electron Microscopy Sciences Supplier No. 7022002
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite.
OAL: 2 3/8"
Diameter: ⅛"
Tip diameter: 0.354"
Tip: 3/16"