Learn More
Pendotech 1/2 Inch Hose Barb Single Use Pressure Sensor
Catalog No. NC0955551
Encompass
This item must be ordered in multiples of 5 per supplier requirements.
Single Use Pressure Sensor, non-sterile, polycarbonate, 1/2 inch hose barb
Catalog No. NC0955551 Supplier Pendotech Supplier No. PRESSN050
This item must be ordered in multiples of 5 per supplier requirements.
Single Use Pressure Sensor, non-sterile, polycarbonate, 1/2 inch hose barb