Learn More
Mopec Press, Shake, and Pour Casting Kit-In-A-Bag
SDP
Catalog No. 22446275 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Evidence Casting Kit
Catalog No. Product Type
22-446-275 Evidence Casting Kit
1 options
Catalog No. 22-446-275 Supplier Mopec Supplier No. FS391
Add to Cart
Add to Cart
A self-contained casting system allows tire and footprint impressions to be cast without separate measuring or mixing. Water and casting material are enclosed inside a sealed bag; breaking the internal bladder and shaking the bag prepares the material for pouring directly into the impression.
Construction
  • Water bladder and casting material in sealed bag
Materials
  • Polyethylene bag
Dimensions
  • 9 W x 11 H in.
  • 6 mil thick outer bag
Performance
  • Place the bag on a flat surface, press down on the bag to break the internal water bladder, shake bag to mix, then pour
Specifications
Product Type Evidence Casting Kit