Learn More
Ace Glass, Inc. Bottle only, pressure, 1850mL, #25, no plug
SDP
Catalog No. NC9028473
Encompass_Preferred
1850mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include #25 plug.
Catalog No. NC9028473 Supplier Ace Glass, Inc. Supplier No. 8648157
1850mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include #25 plug.