Learn More
Ace Glass, Inc. Bottle, pressure, 175mL, 60psi, complete with #25 front-seal plug
SDP
Catalog No. NC9333302
Encompass_Preferred
175mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Includes #25 front-seal plug.
Catalog No. NC9333302 Supplier Ace Glass, Inc. Supplier No. 8648245
175mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Includes #25 front-seal plug.