Learn More
Cole-Parmer Cole-Parmer Pressure Gauge ~ 1/4" NPT (M) process connection; 0 to 5 psi
Catalog No. NC0789440 Shop All Cole-Parmer Products
Encompass_Preferred
Manufactured with a chrome-plated case, these high-quality low-pressure gauges measure inches of water column to 10 psi. Use these gauges to measure low pressure in filter systems, clean rooms, or fluidic systems.
Catalog No. NC0789440 Supplier Cole-Parmer Supplier No. 6835716
Add to Cart
Add to Cart
Manufactured with a chrome-plated case, these high-quality low-pressure gauges measure inches of water column to 10 psi. Use these gauges to measure low pressure in filter systems, clean rooms, or fluidic systems.