Learn More
Waters Corp Pressure Gauge, 1000psi, Bar Dampened
Catalog No. 502391359 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pressure gauge is a replacement part with a 1000psi capacity and a dampened design for stable operation. It is specifically used in SFC80Q, SFC350 Prep, and SFE systems.
  • 1000psi pressure capacity
  • Dampened for stable readings
  • Designed for SFC80Q systems
  • Designed for SFC350 Prep systems
  • Designed for SFE systems
Catalog No. 50-239-1359 Supplier Waters Corp Supplier No. 700006548
This pressure gauge is a replacement part with a 1000psi capacity and a dampened design for stable operation. It is specifically used in SFC80Q, SFC350 Prep, and SFE systems.
  • 1000psi pressure capacity
  • Dampened for stable readings
  • Designed for SFC80Q systems
  • Designed for SFC350 Prep systems
  • Designed for SFE systems