Learn More
Hach Company Pressure Regulator for Synergy Water Systems
Supplier:  Hach Company 2512604
Encompass
Pressure Regulator for the Milli-Q and Synergy Water Systems. Regulates feedwater pressure. Required when feed water is greater than 0.5bar.
Catalog No. 50-199-6151