Learn More
Cole-Parmer Ashcroft 1032S2.0 2" Sanitary Pressure Gauge 60 psi, 3/4" Tri-Clamp
Catalog No. NC1449638 Shop All Cole-Parmer Products
Encompass_Preferred
The Ashcroft Type 1032 Sanitary Pressure Gauge is specifically designed for applications in the food, pharmaceutical and biotechnical industries where small size and sanitary conditions are a priority. This compact autoclavable 2" gauge features all stainless steel construction, temperature-vented case, built-in pressure dampening and a self-draining case to facilitate wash downs. The gauges can also be cleaned or steamed in place.
Catalog No. NC1449638 Supplier Cole-Parmer Supplier No. 6884704
Add to Cart
Add to Cart
The Ashcroft Type 1032 Sanitary Pressure Gauge is specifically designed for applications in the food, pharmaceutical and biotechnical industries where small size and sanitary conditions are a priority. This compact autoclavable 2" gauge features all stainless steel construction, temperature-vented case, built-in pressure dampening and a self-draining case to facilitate wash downs. The gauges can also be cleaned or steamed in place.