Learn More
Valutek™ Pressure Heat Seal Standard Weight Polyester Wiper
Catalog No. 191502003
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
Catalog No. Dimensions (L x W)
19-150-2003 9 x 9 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-2003 Supplier Valutek™ Supplier No. VTPNWPHS99
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed from 100% continuous filament polyester in a double-knit, no-run, interlock pattern with pressure-heat sealed edges. This wiper is an ultraclean and highly absorbent wiper with ultra low levels of particulate and extractable counts.
  • 100% continuous-filament double-knit polyester fiber
  • Pressure-heat sealed edge for lowest available fiber contamination
  • High absorbency and abrasion resistance
  • Excellent chemical compatibility with IPA, Acetone and other common solvents
  • Ultra low levels of particulate and extractable counts
  • Laundered and packaged in the cleanroom, all Valutek™ wipers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 1 -10 (ISO 3-4) critical environment
  • It is designed for critical processing applications where the highest level of contamination control is required
  • It is also commonly used for cleaning medical device products; applying, removing, cleaning and disinfecting solutions; is steam autoclavable for aseptic environments; and used for flat panel, optics cleaning, and chips multi-processing (CMP)
Specifications
Material Polyester
Dimensions (L x W) 9 x 9 in.
Sterility Non-sterile