Learn More
World Precision Instrument Pressure Manometer-SYS-PM100D
SDP
Catalog No. 502538492
Encompass
Hand-held and battery operated, PM Series pressure manometers monitor vac­u­um and pressure in non-corrosive gases. An integral transducer and dig­i­tal display allow easy and accurate pressure readings. Three ver­sions measure pressures in the range of ±1 PSI, ±15 PSI or ±100 PSI. A range switch allows measurement in units of PSI or kPa for the 100 PSI version, and PSI or mmHg for the 15 psi version. Pres­sure can be read on the built-in LCD display or relayed to a chart recorder, os­cil­lo­scope, or computer.
Catalog No. 50-253-8492 Supplier World Precision Instrument Supplier No. SYSPM100D
Hand-held and battery operated, PM Series pressure manometers monitor vac­u­um and pressure in non-corrosive gases. An integral transducer and dig­i­tal display allow easy and accurate pressure readings. Three ver­sions measure pressures in the range of ±1 PSI, ±15 PSI or ±100 PSI. A range switch allows measurement in units of PSI or kPa for the 100 PSI version, and PSI or mmHg for the 15 psi version. Pres­sure can be read on the built-in LCD display or relayed to a chart recorder, os­cil­lo­scope, or computer.