Learn More
Cole-Parmer Fluke 700PMP Hand-Operated Pneumatic Pump, 150 psi
Supplier:  Cole-Parmer 6845074
Encompass
The Fluke 700PMP hand-operated pressure pump provides pressures up to 150 psi. It is a tube type, cylindrical shaped hand pump with a "T" handle at the compressor end and a round knob at the volume adjust end. The pump incorporates a needle valve for venting and a volume adjust vernier for precision adjustment of pressure. Pressure connections are made through one 1/8" NPT internally threaded fitting.
Catalog No. 50-993-406