Learn More
Organomation Pressure Reducing Gas Regulator
SDP
Catalog No. NC2113359
Encompass_Preferred
This regulator steps gas pressure down from 30-160 psig to 0-60 psig. Compatible with all Organomation nitrogen evaporators.
This item is not returnable. View return policy
Catalog No. NC2113359 Supplier Organomation Supplier No. NA0630
This regulator steps gas pressure down from 30-160 psig to 0-60 psig. Compatible with all Organomation nitrogen evaporators.