Learn More
TA Instruments Pressure Regulator USA Only, Purge Gases Thermal Modules, 4000 psi Input 40 psi Output
Catalog No. 5000580745 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Pressure Regulator is for USA use to regulate purge gases for thermal modules, 4000 psi input, 40 psi output, CGA threads suitable for USA only.
Catalog No. 50-005-80745 Supplier TA Instruments Supplier No. 200245.001
Waters TA Instruments Pressure Regulator is for USA use to regulate purge gases for thermal modules, 4000 psi input, 40 psi output, CGA threads suitable for USA only.